siRNA Details
Gene Symbol: PCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271830.1 | NP_001258759.1 | 3 | 203 | ||
NM_174895.2 | NP_777555.1 | 3 | 485 | |||
XM_006722639.2 | XP_006722702.1 | 3 | 705 | |||
XM_017026249.1 | XP_016881738.1 | 5 | 1514 | |||
XM_017026250.1 | XP_016881739.1 | 5 | 1373 | |||
XM_017026251.1 | XP_016881740.1 | 4 | 1329 | |||
XM_017026252.1 | XP_016881741.1 | 3 | 846 | |||
GenBank mRNA | AK130855.1 | - | 3 | 437 | ||
BC025387.1 | - | 3 | 490 | 4824135 | ||
BC038715.1 | - | 3 | 421 | 5268414 | ||
BX281028.1 | - | 3 | 162 | |||
BX647746.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
