siRNA Details
Gene Symbol: DENND6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152678.2 | NP_689891.1 | 20 | 1790 | ||
XM_006713019.3 | XP_006713082.1 | 20 | 1816 | |||
XM_006713020.2 | XP_006713083.1 | 21 | 2022 | |||
XM_017005864.1 | XP_016861353.1 | 21 | 2025 | |||
GenBank mRNA | AK074156.1 | - | 20 | 1777 | ||
AK092288.1 | - | 15 | 1192 | |||
BC040291.1 | - | 20 | 1778 | 4822560 | ||
BC054343.1 | - | 11 | 873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
