siRNA Details
Gene Symbol: INTS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039937.1 | NP_001035026.1 | 19 | 2714 | ||
NM_001306091.1 | NP_001293020.1 | 20 | 3490 | |||
NM_012141.2 | NP_036273.1 | 18 | 3206 | |||
XM_011535040.2 | XP_011533342.1 | 18 | 3303 | |||
XM_011535041.1 | XP_011533343.1 | 17 | 3119 | |||
XM_017020540.1 | XP_016876029.1 | 17 | 2664 | |||
GenBank mRNA | AF097645.1 | - | 18 | 3068 | ||
AK074946.1 | - | 16 | 2296 | |||
AK096696.1 | - | 15 | 2426 | |||
AK128795.1 | - | 16 | 2361 | |||
AK304218.1 | - | 17 | 3008 | |||
AL117626.1 | - | 9 | 1361 | |||
AL833524.1 | - | 19 | 2782 | |||
BC039829.1 | - | 18 | 3080 | 5736353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
