siRNA Details
Gene Symbol: INTS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039937.1 | NP_001035026.1 | 15 | 1958 | ||
NM_001306091.1 | NP_001293020.1 | 16 | 2734 | |||
NM_012141.2 | NP_036273.1 | 14 | 2450 | |||
XM_011535040.2 | XP_011533342.1 | 14 | 2547 | |||
XM_011535041.1 | XP_011533343.1 | 13 | 2363 | |||
XM_017020540.1 | XP_016876029.1 | 13 | 1908 | |||
GenBank mRNA | AF097645.1 | - | 14 | 2312 | ||
AF141326.1 | - | 8 | 1021 | |||
AK074946.1 | - | 12 | 1540 | |||
AK096696.1 | - | 11 | 1670 | |||
AK128795.1 | - | 12 | 1605 | |||
AK304218.1 | - | 13 | 2252 | |||
AL117626.1 | - | 5 | 605 | |||
AL833524.1 | - | 15 | 2026 | |||
BC013358.2 | - | 14 | 2328 | |||
BC039829.1 | - | 14 | 2324 | 5736353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
