siRNA Details
Gene Symbol: INTS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039937.1 | NP_001035026.1 | 17 | 2254 | ||
NM_001306091.1 | NP_001293020.1 | 18 | 3030 | |||
NM_012141.2 | NP_036273.1 | 16 | 2746 | |||
XM_011535040.2 | XP_011533342.1 | 16 | 2843 | |||
XM_011535041.1 | XP_011533343.1 | 15 | 2659 | |||
XM_017020540.1 | XP_016876029.1 | 15 | 2204 | |||
GenBank mRNA | AF097645.1 | - | 16 | 2608 | ||
AF141326.1 | - | 10 | 1317 | |||
AK074946.1 | - | 14 | 1836 | |||
AK096696.1 | - | 13 | 1966 | |||
AK128795.1 | - | 14 | 1901 | |||
AK296801.1 | - | 9 | 1429 | |||
AK304218.1 | - | 15 | 2548 | |||
AL117626.1 | - | 7 | 901 | |||
AL833524.1 | - | 17 | 2322 | |||
BC013358.2 | - | 16 | 2624 | |||
BC039829.1 | - | 16 | 2620 | 5736353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
