siRNA Details
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181725.3 | NP_859076.3 | 3 | 380 | ||
XM_017024572.1 | XP_016880061.1 | 3 | 368 | |||
GenBank mRNA | AF161385.1 | - | 328 | |||
AK022822.1 | - | 3 | 357 | |||
BC006985.1 | - | 2 | 582 | 3930328 | ||
BF975211.1 | - | 3 | 353 | |||
BP255803.1 | - | 3 | 381 | |||
DA027807.1 | - | 3 | 368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
