siRNA Details
Gene Symbol: EDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302455.1 | NP_001289384.1 | 5 | 1438 | ||
NM_001302456.1 | NP_001289385.1 | 4 | 1388 | |||
NM_207032.2 | NP_996915.1 | 5 | 1469 | |||
NM_207033.2 | NP_996916.1 | 4 | 1392 | |||
NM_207034.2 | NP_996917.1 | 5 | 1434 | |||
XM_005260312.4 | XP_005260369.1 | 5 | 1483 | |||
XM_005260313.4 | XP_005260370.1 | 5 | 1460 | |||
XM_006723734.3 | XP_006723797.1 | 5 | 1510 | |||
XM_011528655.2 | XP_011526957.1 | 5 | 1463 | |||
GenBank mRNA | BC008876.2 | - | 5 | 1434 | 4139312 | |
BC053866.1 | - | 5 | 1453 | 5202210 | ||
J05081.1 | - | 6 | 1021 | |||
X52001.1 | - | 4 | 1116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
