siRNA Details
Gene Symbol: EDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302455.1 | NP_001289384.1 | 5 | 992 | ||
NM_001302456.1 | NP_001289385.1 | 4 | 942 | |||
NM_207032.2 | NP_996915.1 | 5 | 1023 | |||
NM_207033.2 | NP_996916.1 | 4 | 946 | |||
NM_207034.2 | NP_996917.1 | 5 | 988 | |||
XM_005260312.4 | XP_005260369.1 | 5 | 1037 | |||
XM_005260313.4 | XP_005260370.1 | 5 | 1014 | |||
XM_006723734.3 | XP_006723797.1 | 5 | 1064 | |||
XM_011528655.2 | XP_011526957.1 | 5 | 1017 | |||
GenBank mRNA | BC008876.2 | - | 5 | 988 | 4139312 | |
BC053866.1 | - | 5 | 1007 | 5202210 | ||
BT007085.1 | - | 5 | 602 | |||
DQ096281.1 | - | 4 | 556 | |||
J05081.1 | - | 5 | 795 | |||
S50864.1 | - | 3 | 286 | |||
S50866.1 | - | 2 | 205 | |||
X52001.1 | - | 4 | 670 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
