siRNA Details
Gene Symbol: UBQLNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145053.4 | NP_659490.4 | 1 | 378 | ||
GenBank mRNA | AK057255.1 | - | 1 | 381 | ||
BC012183.1 | - | 1 | 238 | 4554912 | ||
BC034977.2 | - | 1 | 347 | 4822902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
