siRNA Details
Gene Symbol: FAM47A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203408.3 | NP_981953.2 | 1 | 1376 | ||
GenBank mRNA | AK292354.1 | - | 1 | 1360 | ||
BC026171.2 | - | 1 | 1360 | 4827969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
