siRNA Details
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185060.1 | NP_001171989.1 | 5 | 1270 | ||
NM_001185061.1 | NP_001171990.1 | 4 | 1445 | |||
NM_001185062.1 | NP_001171991.1 | 4 | 1217 | |||
NM_198098.2 | NP_932766.1 | 4 | 1590 | |||
GenBank mRNA | BC022486.1 | - | 4 | 1537 | 4799283 | |
BC141905.1 | - | 1 | 137 | |||
U41517.1 | - | 4 | 1512 | |||
U41518.1 | - | 4 | 1130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
