siRNA Details
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185060.1 | NP_001171989.1 | 5 | 829 | ||
NM_001185061.1 | NP_001171990.1 | 4 | 1004 | |||
NM_001185062.1 | NP_001171991.1 | 4 | 776 | |||
NM_198098.2 | NP_932766.1 | 4 | 1149 | |||
GenBank mRNA | AK298313.1 | - | 5 | 910 | ||
AK300995.1 | - | 5 | 961 | |||
AK301351.1 | - | 4 | 757 | |||
AK309608.1 | - | 5 | 829 | |||
AK309691.1 | - | 4 | 848 | |||
AY312580.1 | - | 3 | 258 | |||
BC022486.1 | - | 4 | 1096 | 4799283 | ||
M77829.1 | - | 4 | 1077 | |||
S73482.1 | - | 4 | 1077 | |||
U41517.1 | - | 4 | 1072 | |||
U41518.1 | - | 4 | 690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
