siRNA Details
Gene Symbol: ZNF507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136156.1 | NP_001129628.1 | 3 | 937 | ||
NM_014910.4 | NP_055725.2 | 2 | 843 | |||
GenBank mRNA | AB029007.1 | - | 3 | 898 | ||
AK292526.1 | - | 2 | 841 | |||
AL713775.1 | - | 3 | 824 | |||
BC136417.1 | - | 3 | 873 | 9020405 | ||
BX640881.1 | - | 2 | 805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
