siRNA Details
Gene Symbol: CKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008938.3 | NP_001008938.1 | 44 | 6133 | ||
NM_014756.3 | NP_055571.2 | 43 | 5953 | |||
GenBank mRNA | BC035554.1 | - | 19 | 2916 | ||
BC120869.1 | - | 43 | 6027 | 40072423 | ||
BC120870.1 | - | 43 | 6046 | 40072425 | ||
D43948.1 | - | 43 | 6054 | |||
X92474.1 | - | 42 | 5853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
