siRNA Details
Gene Symbol: OR10G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005466.2 | NP_001005466.2 | 1 | 1020 | ||
GenBank mRNA | BC137371.1 | - | 1 | 1020 | 9021369 | |
BC137382.1 | - | 1 | 1020 | 9021380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
