siRNA Details
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025092.4 | NP_079368.3 | 11, 12 | 1824 | ||
XM_011520383.2 | XP_011518685.1 | 14, 15 | 2750 | |||
XM_011520384.2 | XP_011518686.1 | 13, 14 | 2861 | |||
XM_017018355.1 | XP_016873844.1 | 12, 13 | 2202 | |||
GenBank mRNA | AK074155.1 | - | 8, 9 | 2986 | ||
AK090428.1 | - | 5, 6 | 1441 | |||
AK296137.1 | - | 7, 8 | 703 | |||
AL832932.1 | - | 4, 5 | 482 | |||
BC048336.1 | - | 2, 3 | 371 | |||
BC066641.1 | - | 10, 11 | 1531 | |||
BC109257.2 | - | 8, 9 | 1223 | 40024867 | ||
BC109258.2 | - | 8, 9 | 1223 | 40024874 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
