siRNA Details
Gene Symbol: KRT25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181534.3 | NP_853512.1 | 8 | 1641 | ||
XM_011524414.1 | XP_011522716.1 | 9 | 1629 | |||
GenBank mRNA | AJ564204.1 | - | 8 | 1622 | ||
AK129503.1 | - | 8 | 1443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
