siRNA Details
Gene Symbol: FGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291812.1 | NP_001278741.1 | 3 | 1717 | ||
NM_004464.3 | NP_004455.2 | 3 | 2005 | |||
NM_033143.2 | NP_149134.1 | 2 | 1901 | |||
GenBank mRNA | AF171928.1 | - | 2 | 1814 | ||
AK090557.1 | - | 3 | 1717 | |||
BC131502.1 | - | 3 | 1906 | 40082281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
