siRNA Details
Gene Symbol: CYP4A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010969.3 | NP_001010969.2 | 2 | 279 | ||
NM_001308102.1 | NP_001295031.1 | 2 | 279 | |||
XM_005270767.3 | XP_005270824.1 | 2 | 261 | |||
XM_005270770.3 | XP_005270827.1 | 2 | 261 | |||
GenBank mRNA | AY280371.1 | - | 2 | 269 | ||
AY280372.1 | - | 2 | 218 | |||
BC142607.1 | - | 2 | 248 | 40147121 | ||
BC142690.1 | - | 2 | 250 | 40148647 | ||
BC148248.1 | - | 2 | 279 | 40132876 |
Gene Symbol: CYP4A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000778.3 | NP_000769.2 | 2 | 269 | ||
NM_001319155.1 | NP_001306084.1 | 2 | 269 | |||
NR_134988.1 | 2 | 269 | ||||
NR_134989.1 | 2 | 269 | ||||
NR_134990.1 | 2 | 269 | ||||
NR_134991.1 | 2 | 269 | ||||
NR_134992.1 | 2 | 269 | ||||
NR_134993.1 | 2 | 269 | ||||
NR_134994.1 | 2 | 269 | ||||
XM_005270539.1 | XP_005270596.1 | 2 | 261 | |||
XM_017000464.1 | XP_016855953.1 | 2 | 261 | |||
XM_017000465.1 | XP_016855954.1 | 2 | 188 | |||
GenBank mRNA | AK298267.1 | - | 2 | 210 | ||
AK298413.1 | - | 2 | 210 | |||
BC022851.1 | - | 2 | 265 | |||
BC041158.1 | - | 2 | 249 | 5183289 | ||
CD013974.1 | - | 2 | 269 | |||
CD013975.1 | - | 2 | 269 | |||
CD013976.1 | - | 2 | 269 | |||
CD013977.1 | - | 2 | 269 | |||
CD013978.1 | - | 2 | 269 | |||
CD013979.1 | - | 2 | 269 | |||
CD013980.1 | - | 2 | 269 | |||
CD014132.1 | - | 2 | 269 | |||
D13705.1 | - | 2 | 241 | |||
D26481.1 | - | 2 | 233 | |||
L04751.1 | - | 2 | 250 | |||
S67580.1 | - | 2 | 259 | |||
S67581.1 | - | 2 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
