siRNA Details
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014453.3 | NP_055268.1 | 6 | 857 | ||
NM_198426.2 | NP_940818.1 | 6 | 859 | |||
XM_005258746.2 | XP_005258803.1 | 5 | 1486 | |||
XM_005258747.3 | XP_005258804.1 | 5 | 1609 | |||
GenBank mRNA | AF042384.1 | - | 6 | 825 | ||
BC002502.2 | - | 6 | 829 | 3051024 | ||
BU147101.1 | - | 4 | 504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
