siRNA Details
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014453.3 | NP_055268.1 | 3 | 442 | ||
NM_198426.2 | NP_940818.1 | 3 | 444 | |||
XM_005258746.2 | XP_005258803.1 | 3 | 942 | |||
XM_005258747.3 | XP_005258804.1 | 3 | 1065 | |||
GenBank mRNA | AF042384.1 | - | 3 | 410 | ||
AK311974.1 | - | 3 | 330 | |||
AY364248.1 | - | 2 | 281 | |||
BC002502.2 | - | 3 | 414 | 3051024 | ||
BG546479.1 | - | 3 | 438 | |||
BI457978.1 | - | 3 | 434 | |||
BT007298.1 | - | 2 | 281 | |||
BU147101.1 | - | 1 | 89 | |||
CN305066.1 | - | 3 | 445 | |||
CR457002.1 | - | 2 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
