siRNA Details
Gene Symbol: PTAFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164721.1 | NP_001158193.1 | 1 | 192 | ||
NM_001164722.2 | NP_001158194.1 | 1 | 192 | |||
GenBank mRNA | AK292053.1 | - | 1 | 192 | ||
DA705644.1 | - | 1 | 182 | |||
S52624.1 | - | 1 | 130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
