siRNA Details
Gene Symbol: OR52I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005170.2 | NP_001005170.1 | 1 | 761 | ||
GenBank mRNA | BC136921.1 | - | 1 | 761 | 9020912 | |
BC151144.1 | - | 1 | 761 | 9020913 |
Gene Symbol: OR52I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005169.1 | NP_001005169.1 | 1 | 661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
