siRNA Details
Gene Symbol: YPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005404.3 | NP_001005404.1 | 5 | 697 | ||
XM_017024621.1 | XP_016880110.1 | 5 | 715 | |||
GenBank mRNA | AB098736.1 | - | 5 | 620 | ||
AF305195.1 | - | 4 | 513 | |||
AI540782.1 | - | 4 | 388 | |||
BC127924.1 | - | 4 | 428 | 40132521 | ||
BC132884.1 | - | 5 | 687 | 40146906 | ||
BC132886.1 | - | 5 | 687 | 40146908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
