siRNA Details
Gene Symbol: C1orf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010881.1 | NP_001010881.1 | 9 | 2257 | ||
XM_011541269.2 | XP_011539571.2 | 10 | 2504 | |||
XM_011541271.2 | XP_011539573.2 | 10 | 2299 | |||
XM_011541272.2 | XP_011539574.1 | 10 | 2353 | |||
XM_011541274.2 | XP_011539576.2 | 9 | 2371 | |||
XM_011541275.2 | XP_011539577.2 | 9 | 2168 | |||
XM_011541276.2 | XP_011539578.1 | 10 | 2353 | |||
XM_011541277.2 | XP_011539579.1 | 10 | 2353 | |||
XM_011541278.2 | XP_011539580.1 | 10 | 2353 | |||
XM_011541280.2 | XP_011539582.2 | 5 | 604 | |||
XM_011541281.2 | XP_011539583.2 | 5 | 585 | |||
XM_017001062.1 | XP_016856551.1 | 11 | 2491 | |||
XM_017001063.1 | XP_016856552.1 | 10 | 2344 | |||
XM_017001064.1 | XP_016856553.1 | 10 | 2353 | |||
GenBank mRNA | AL833920.1 | - | 7 | 1119 | ||
AL834308.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
