siRNA Details
Gene Symbol: KCNRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173605.1 | NP_775876.1 | 1 | 469 | ||
NM_199464.2 | NP_955751.1 | 1 | 469 | |||
GenBank mRNA | AK313401.1 | - | 1 | 320 | ||
AY129653.1 | - | 1 | 279 | |||
AY129654.1 | - | 1 | 287 | |||
AY169387.1 | - | 1 | 469 | |||
AY169388.1 | - | 1 | 469 | |||
AY190921.1 | - | 1 | 249 | |||
AY190922.1 | - | 1 | 249 | |||
BC020887.1 | - | 1 | 279 | 4690662 | ||
BC032343.1 | - | 1 | 263 | 5207702 |
Gene Symbol: TRIM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007278.2 | NP_001007279.1 | 4 | 4209 | ||
NM_005798.4 | NP_005789.2 | 3 | 4010 | |||
NM_052811.3 | NP_434698.1 | 3 | 4095 | |||
NM_213590.2 | NP_998755.1 | 2 | 4545 | |||
GenBank mRNA | AK056841.1 | - | 1 | 21 | ||
AL832695.1 | - | 1 | 239 | |||
AY455758.1 | - | 2 | 4520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
