siRNA Details
Gene Symbol: KCNRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173605.1 | NP_775876.1 | 1 | 531 | ||
NM_199464.2 | NP_955751.1 | 1 | 531 | |||
GenBank mRNA | AK313401.1 | - | 1 | 382 | ||
AY129653.1 | - | 1 | 341 | |||
AY129654.1 | - | 1 | 349 | |||
AY169387.1 | - | 1 | 531 | |||
AY169388.1 | - | 1 | 531 | |||
AY190921.1 | - | 1 | 311 | |||
AY190922.1 | - | 1 | 311 | |||
BC020887.1 | - | 1 | 341 | 4690662 | ||
BC032343.1 | - | 1 | 325 | 5207702 |
Gene Symbol: TRIM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007278.2 | NP_001007279.1 | 4 | 4271 | ||
NM_005798.4 | NP_005789.2 | 3 | 4072 | |||
NM_052811.3 | NP_434698.1 | 3 | 4157 | |||
NM_213590.2 | NP_998755.1 | 2 | 4607 | |||
GenBank mRNA | AK056841.1 | - | 1 | 83 | ||
AL832695.1 | - | 1 | 301 | |||
AY455758.1 | - | 2 | 4582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
