siRNA Details
Gene Symbol: KCNRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173605.1 | NP_775876.1 | 1 | 345 | ||
NM_199464.2 | NP_955751.1 | 1 | 345 | |||
GenBank mRNA | AK313401.1 | - | 1 | 196 | ||
AY129653.1 | - | 1 | 155 | |||
AY129654.1 | - | 1 | 163 | |||
AY169387.1 | - | 1 | 345 | |||
AY169388.1 | - | 1 | 345 | |||
AY190921.1 | - | 1 | 125 | |||
AY190922.1 | - | 1 | 125 | |||
BC020887.1 | - | 1 | 155 | 4690662 | ||
BC032343.1 | - | 1 | 139 | 5207702 |
Gene Symbol: TRIM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007278.2 | NP_001007279.1 | 4 | 4085 | ||
NM_005798.4 | NP_005789.2 | 3 | 3886 | |||
NM_052811.3 | NP_434698.1 | 3 | 3971 | |||
NM_213590.2 | NP_998755.1 | 2 | 4421 | |||
GenBank mRNA | AL832695.1 | - | 1 | 115 | ||
AY455758.1 | - | 2 | 4396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
