siRNA Details
Gene Symbol: SHOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163678.1 | NP_001157150.1 | 3, 4 | 751 | ||
NM_003030.4 | NP_003021.3 | 4, 5 | 823 | |||
NM_006884.3 | NP_006875.2 | 3, 4 | 751 | |||
XM_006713727.3 | XP_006713790.1 | 4, 5 | 872 | |||
XM_006713728.3 | XP_006713791.1 | 4, 5 | 797 | |||
XM_017007053.1 | XP_016862542.1 | 4, 5 | 1419 | |||
XM_017007054.1 | XP_016862543.1 | 5, 6 | 1266 | |||
XM_017007055.1 | XP_016862544.1 | 4, 5 | 469 | |||
GenBank mRNA | AF022654.1 | - | 2, 3 | 422 | ||
AJ002367.1 | - | 3, 4 | 638 | |||
AJ002368.1 | - | 2, 3 | 267 | |||
AK095338.1 | - | 3, 4 | 1760 | |||
BC008829.2 | - | 4, 5 | 823 | 4122010 | ||
DA447204.1 | - | 3, 4 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
