siRNA Details
Gene Symbol: SHOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163678.1 | NP_001157150.1 | 2 | 557 | ||
NM_003030.4 | NP_003021.3 | 3 | 629 | |||
NM_006884.3 | NP_006875.2 | 2 | 557 | |||
XM_006713727.3 | XP_006713790.1 | 3 | 678 | |||
XM_006713728.3 | XP_006713791.1 | 3 | 603 | |||
XM_017007053.1 | XP_016862542.1 | 3 | 1225 | |||
XM_017007054.1 | XP_016862543.1 | 4 | 1072 | |||
XM_017007055.1 | XP_016862544.1 | 3 | 275 | |||
GenBank mRNA | AF022654.1 | - | 1 | 228 | ||
AJ002367.1 | - | 2 | 444 | |||
AJ002368.1 | - | 1 | 73 | |||
AK095338.1 | - | 2 | 1566 | |||
BC008829.2 | - | 3 | 629 | 4122010 | ||
DA447204.1 | - | 2 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
