siRNA Details
Gene Symbol: CYFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033028.1 | NP_001028200.1 | 6 | 1296 | ||
NM_001287810.2 | NP_001274739.1 | 22 | 2567 | |||
NM_001324119.1 | NP_001311048.1 | 21 | 2396 | |||
NM_001324120.1 | NP_001311049.1 | 21 | 2575 | |||
NM_001324122.1 | NP_001311051.1 | 22 | 2596 | |||
NM_001324123.1 | NP_001311052.1 | 21 | 2475 | |||
NM_001324124.1 | NP_001311053.1 | 20 | 2358 | |||
NM_001324125.1 | NP_001311054.1 | 18 | 2157 | |||
NM_001324126.1 | NP_001311055.1 | 20 | 2346 | |||
NM_014608.4 | NP_055423.1 | 21 | 2448 | |||
XM_011543873.2 | XP_011542175.1 | 21 | 2797 | |||
XM_011543874.1 | XP_011542176.1 | 21 | 2830 | |||
XM_011543876.2 | XP_011542178.2 | 21 | 2398 | |||
XM_017022023.1 | XP_016877512.1 | 21 | 2803 | |||
XM_017022024.1 | XP_016877513.1 | 21 | 2811 | |||
GenBank mRNA | AK125936.1 | - | 20 | 2501 | ||
AK127094.1 | - | 6 | 1296 | |||
AK291604.1 | - | 21 | 2375 | |||
AL357618.1 | - | 5 | 440 | |||
AY763577.1 | - | 21 | 2412 | |||
AY763578.1 | - | 22 | 2478 | |||
AY763579.1 | - | 6 | 1296 | |||
BC001306.2 | - | 4 | 264 | |||
BC005097.1 | - | 21 | 2339 | 3163591 | ||
BC063869.1 | - | 6 | 1090 | |||
D38549.1 | - | 21 | 2342 | |||
KJ534819.1 | - | 20 | 2287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
