siRNA Details
Gene Symbol: CYFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033028.1 | NP_001028200.1 | 5 | 1191 | ||
NM_001287810.2 | NP_001274739.1 | 21 | 2462 | |||
NM_001324119.1 | NP_001311048.1 | 20 | 2291 | |||
NM_001324120.1 | NP_001311049.1 | 20 | 2470 | |||
NM_001324122.1 | NP_001311051.1 | 21 | 2491 | |||
NM_001324123.1 | NP_001311052.1 | 20 | 2370 | |||
NM_001324124.1 | NP_001311053.1 | 19 | 2253 | |||
NM_001324125.1 | NP_001311054.1 | 17 | 2052 | |||
NM_001324126.1 | NP_001311055.1 | 19 | 2241 | |||
NM_014608.4 | NP_055423.1 | 20 | 2343 | |||
XM_011543873.2 | XP_011542175.1 | 20 | 2692 | |||
XM_011543874.1 | XP_011542176.1 | 20 | 2725 | |||
XM_011543876.2 | XP_011542178.2 | 20 | 2293 | |||
XM_017022023.1 | XP_016877512.1 | 20 | 2698 | |||
XM_017022024.1 | XP_016877513.1 | 20 | 2706 | |||
GenBank mRNA | AK125936.1 | - | 19 | 2396 | ||
AK127094.1 | - | 5 | 1191 | |||
AK291604.1 | - | 20 | 2270 | |||
AL357618.1 | - | 4 | 335 | |||
AY763577.1 | - | 20 | 2307 | |||
AY763578.1 | - | 21 | 2373 | |||
AY763579.1 | - | 5 | 1191 | |||
BC001306.2 | - | 3 | 159 | |||
BC005097.1 | - | 20 | 2234 | 3163591 | ||
BC063869.1 | - | 5 | 985 | |||
D38549.1 | - | 20 | 2237 | |||
KJ534819.1 | - | 19 | 2182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
