siRNA Details
Gene Symbol: CYFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033028.1 | NP_001028200.1 | 14 | 2395 | ||
NM_001287810.2 | NP_001274739.1 | 30 | 3666 | |||
NM_001324119.1 | NP_001311048.1 | 29 | 3495 | |||
NM_001324120.1 | NP_001311049.1 | 29 | 3674 | |||
NM_001324122.1 | NP_001311051.1 | 30 | 3695 | |||
NM_001324123.1 | NP_001311052.1 | 29 | 3574 | |||
NM_001324124.1 | NP_001311053.1 | 28 | 3457 | |||
NM_001324125.1 | NP_001311054.1 | 26 | 3256 | |||
NM_001324126.1 | NP_001311055.1 | 28 | 3445 | |||
NM_014608.4 | NP_055423.1 | 29 | 3547 | |||
XM_011543873.2 | XP_011542175.1 | 29 | 3896 | |||
XM_011543874.1 | XP_011542176.1 | 29 | 3929 | |||
XM_011543876.2 | XP_011542178.2 | 29 | 3497 | |||
XM_017022023.1 | XP_016877512.1 | 29 | 3902 | |||
XM_017022024.1 | XP_016877513.1 | 29 | 3910 | |||
GenBank mRNA | AK125784.1 | - | 8 | 2533 | ||
AK125936.1 | - | 28 | 3522 | |||
AK127094.1 | - | 14 | 2395 | |||
AK291604.1 | - | 29 | 3474 | |||
AY763577.1 | - | 29 | 3511 | |||
AY763578.1 | - | 30 | 3577 | |||
AY763579.1 | - | 14 | 2395 | |||
AY763580.1 | - | 8 | 2533 | |||
BC001306.2 | - | 12 | 1363 | |||
BC005097.1 | - | 29 | 3438 | 3163591 | ||
BC063869.1 | - | 14 | 2189 | |||
D38549.1 | - | 29 | 3441 | |||
KJ534819.1 | - | 28 | 3386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
