siRNA Details
Gene Symbol: RHBDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005498.3 | NP_001005498.2 | 9 | 1399 | ||
NM_024599.5 | NP_078875.4 | 10 | 1561 | |||
XM_005257669.3 | XP_005257726.1 | 10 | 1922 | |||
XM_005257670.1 | XP_005257727.1 | 10 | 1431 | |||
XM_011525249.2 | XP_011523551.1 | 10 | 2878 | |||
XM_011525250.2 | XP_011523552.1 | 11 | 1588 | |||
XM_011525251.2 | XP_011523553.1 | 10 | 1405 | |||
XM_017025079.1 | XP_016880568.1 | 10 | 1835 | |||
GenBank mRNA | AK025994.1 | - | 5 | 585 | ||
AK074034.1 | - | 8 | 1866 | |||
AK225684.1 | - | 5 | 538 | |||
AK292135.1 | - | 10 | 1554 | |||
AK292166.1 | - | 9 | 1354 | |||
AK303434.1 | - | 8 | 1215 | |||
AL833680.1 | - | 4 | 454 | |||
AY500247.1 | - | 9 | 1250 | |||
BC016034.1 | - | 10 | 1541 | 3910543 | ||
BC035829.1 | - | 5 | 647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
