siRNA Details
Gene Symbol: SERTAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014755.2 | NP_055570.1 | 2 | 4357 | ||
XM_005264669.2 | XP_005264726.1 | 2 | 4875 | |||
XM_011533204.1 | XP_011531506.1 | 2 | 4094 | |||
XM_011533205.2 | XP_011531507.1 | 2 | 4411 | |||
GenBank mRNA | D50917.1 | - | 2 | 4356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
