siRNA Details
Gene Symbol: CHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316690.1 | NP_001303619.1 | 1 | 1441 | ||
NM_017780.3 | NP_060250.2 | 2 | 1759 | |||
XM_011517553.2 | XP_011515855.1 | 2 | 1780 | |||
XM_011517554.2 | XP_011515856.1 | 2 | 1758 | |||
XM_011517555.2 | XP_011515857.1 | 2 | 1780 | |||
XM_011517560.2 | XP_011515862.1 | 2 | 1780 | |||
XM_017013612.1 | XP_016869101.1 | 2 | 1834 | |||
XM_017013613.1 | XP_016869102.1 | 2 | 1780 | |||
GenBank mRNA | AK123158.1 | - | 1 | 550 | ||
BC014681.1 | - | 2 | 1654 | |||
BC053890.1 | - | 2 | 1724 | |||
BC068000.1 | - | 2 | 1744 | |||
BC080627.1 | - | 1 | 1340 | |||
BC110818.1 | - | 2 | 1716 | |||
GU060498.1 | - | 1 | 1335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
