siRNA Details
Gene Symbol: OR5K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004737.1 | NP_001004737.1 | 1 | 740 | ||
GenBank mRNA | BC136724.1 | - | 1 | 817 | 9020714 | |
BC136725.1 | - | 1 | 817 | 9020715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
