siRNA Details
Gene Symbol: RNF216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207111.3 | NP_996994.1 | 15 | 2534 | ||
NM_207116.2 | NP_996999.1 | 15 | 2363 | |||
XM_005249785.2 | XP_005249842.1 | 17 | 2854 | |||
XM_011515436.1 | XP_011513738.1 | 13 | 2273 | |||
XM_017012363.1 | XP_016867852.1 | 15 | 2218 | |||
XM_017012365.1 | XP_016867854.1 | 14 | 2176 | |||
GenBank mRNA | AF228527.1 | - | 5 | 580 | ||
AF513717.1 | - | 15 | 2199 | |||
AF513718.1 | - | 15 | 2370 | |||
AK000916.1 | - | 7 | 758 | |||
AK024787.1 | - | 1 | 106 | |||
AK123635.1 | - | 12 | 1688 | |||
AK292396.1 | - | 15 | 2543 | |||
AK297416.1 | - | 16 | 1718 | |||
AK314332.1 | - | 12 | 1315 | |||
AY062174.1 | - | 12 | 1380 | |||
AY177396.1 | - | 12 | 1438 | |||
AY177397.1 | - | 14 | 2448 | |||
AY177398.1 | - | 14 | 2452 | |||
BC000787.2 | - | 9 | 948 | |||
BC063825.1 | - | 15 | 2273 | 6504476 | ||
BX537406.1 | - | 15 | 2295 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_080561, NM_207110, AY572785, AY572786, BC042205, and others.
Gene Ontology Categories:
