siRNA Details
Gene Symbol: RNF216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207111.3 | NP_996994.1 | 14 | 2408 | ||
NM_207116.2 | NP_996999.1 | 14 | 2237 | |||
XM_005249785.2 | XP_005249842.1 | 16 | 2728 | |||
XM_011515436.1 | XP_011513738.1 | 12 | 2147 | |||
XM_017012363.1 | XP_016867852.1 | 14 | 2092 | |||
XM_017012364.1 | XP_016867853.1 | 14 | 2381 | |||
XM_017012365.1 | XP_016867854.1 | 13 | 2050 | |||
GenBank mRNA | AF228527.1 | - | 4 | 454 | ||
AF513717.1 | - | 14 | 2073 | |||
AF513718.1 | - | 14 | 2244 | |||
AK000916.1 | - | 6 | 632 | |||
AK092767.1 | - | 14 | 2323 | |||
AK123635.1 | - | 11 | 1562 | |||
AK292396.1 | - | 14 | 2417 | |||
AK297416.1 | - | 15 | 1592 | |||
AK314332.1 | - | 11 | 1189 | |||
AY062174.1 | - | 11 | 1254 | |||
AY177396.1 | - | 11 | 1312 | |||
AY177397.1 | - | 13 | 2322 | |||
AY177398.1 | - | 13 | 2326 | |||
BC000787.2 | - | 8 | 822 | |||
BC063825.1 | - | 14 | 2147 | 6504476 | ||
BX537406.1 | - | 14 | 2169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
