siRNA Details
Gene Symbol: RNF216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207111.3 | NP_996994.1 | 15, 16 | 2639 | ||
NM_207116.2 | NP_996999.1 | 15, 16 | 2468 | |||
XM_005249785.2 | XP_005249842.1 | 17, 18 | 2959 | |||
XM_011515436.1 | XP_011513738.1 | 13, 14 | 2378 | |||
XM_017012363.1 | XP_016867852.1 | 15, 16 | 2323 | |||
XM_017012365.1 | XP_016867854.1 | 14, 15 | 2281 | |||
GenBank mRNA | AF228527.1 | - | 5, 6 | 685 | ||
AF513717.1 | - | 15, 16 | 2304 | |||
AF513718.1 | - | 15, 16 | 2475 | |||
AK000916.1 | - | 7, 8 | 863 | |||
AK024787.1 | - | 1, 2 | 211 | |||
AK123635.1 | - | 12, 13 | 1793 | |||
AK292396.1 | - | 15, 16 | 2648 | |||
AK297231.1 | - | 6, 7 | 593 | |||
AK297416.1 | - | 16, 17 | 1823 | |||
AK314332.1 | - | 12, 13 | 1420 | |||
AY062174.1 | - | 12, 13 | 1485 | |||
AY177396.1 | - | 12, 13 | 1543 | |||
AY177397.1 | - | 14, 15 | 2553 | |||
AY177398.1 | - | 14, 15 | 2557 | |||
BC000787.2 | - | 9, 10 | 1053 | |||
BC063825.1 | - | 15, 16 | 2378 | 6504476 | ||
BX537406.1 | - | 15, 16 | 2400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
