siRNA Details
Gene Symbol: EPSTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002264.2 | NP_001002264.1 | 13 | 1556 | ||
NM_033255.3 | NP_150280.1 | 11 | 1491 | |||
XM_005266596.1 | XP_005266653.1 | 12 | 1523 | |||
XM_005266597.1 | XP_005266654.1 | 12 | 1524 | |||
XM_011535312.1 | XP_011533614.1 | 13 | 2766 | |||
XM_011535313.1 | XP_011533615.1 | 12 | 1461 | |||
XM_011535314.1 | XP_011533616.1 | 8 | 958 | |||
XM_017020853.1 | XP_016876342.1 | 13 | 1346 | |||
XM_017020854.1 | XP_016876343.1 | 14 | 1932 | |||
XM_017020855.1 | XP_016876344.1 | 12 | 2145 | |||
GenBank mRNA | AF396928.1 | - | 11 | 1450 | ||
AK292461.1 | - | 11 | 1461 | |||
AL831953.1 | - | 11 | 2916 | |||
BC023660.2 | - | 13 | 1516 | 4865634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
