siRNA Details
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285389.2 | NP_001272318.1 | 2 | 70 | ||
NM_001285391.2 | NP_001272320.2 | 2 | 393 | |||
NM_001285392.2 | NP_001272321.1 | 2 | 187 | |||
NM_001285393.2 | NP_001272322.1 | 2 | 124 | |||
NM_004414.6 | NP_004405.3 | 2 | 396 | |||
NM_203417.2 | NP_981962.1 | 2 | 182 | |||
NM_203418.2 | NP_981963.1 | 2 | 361 | |||
XM_005260929.1 | XP_005260986.1 | 2 | 317 | |||
XM_005260930.1 | XP_005260987.1 | 2 | 305 | |||
XM_017028282.1 | XP_016883771.1 | 3 | 722 | |||
XM_017028283.1 | XP_016883772.1 | 3 | 722 | |||
GenBank mRNA | AF400429.1 | - | 2 | 54 | ||
AK055845.1 | - | 1 | 593 | |||
AK092184.1 | - | 2 | 317 | |||
AK131569.1 | - | 2 | 314 | |||
AK293293.1 | - | 2 | 175 | |||
AK299444.1 | - | 2 | 111 | |||
AL556803.3 | - | 2 | 396 | |||
AY325903.1 | - | 2 | 388 | |||
BC002864.2 | - | 2 | 156 | 3944959 | ||
BT007363.1 | - | 2 | 100 | |||
BX419260.2 | - | 2 | 314 | |||
CR456878.1 | - | 1 | 22 | |||
CR542065.1 | - | 2 | 100 | |||
DA503835.1 | - | 3 | 277 | |||
DA586027.1 | - | 2 | 124 | |||
DB023862.1 | - | 2 | 187 | |||
DC331441.1 | - | 2 | 361 | |||
U28833.2 | - | 2 | 70 | |||
U85265.2 | - | 2 | 181 | |||
U85266.2 | - | 2 | 124 | |||
U85267.2 | - | 2 | 243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
