siRNA Details
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285389.2 | NP_001272318.1 | 4 | 1625 | ||
NM_001285391.2 | NP_001272320.2 | 4 | 1948 | |||
NM_001285392.2 | NP_001272321.1 | 4 | 1742 | |||
NM_001285393.2 | NP_001272322.1 | 4 | 1679 | |||
NM_004414.6 | NP_004405.3 | 4 | 1951 | |||
NM_203417.2 | NP_981962.1 | 4 | 1737 | |||
NM_203418.2 | NP_981963.1 | 4 | 1916 | |||
XM_017028283.1 | XP_016883772.1 | 5 | 2277 | |||
GenBank mRNA | AK055845.1 | - | 3 | 2148 | ||
AK092184.1 | - | 4 | 1872 | |||
AK131569.1 | - | 4 | 1869 | |||
AY325903.1 | - | 4 | 1943 | |||
BC002864.2 | - | 4 | 1711 | 3944959 | ||
U28833.2 | - | 4 | 1625 | |||
U85265.2 | - | 4 | 1736 | |||
U85266.2 | - | 4 | 1679 | |||
U85267.2 | - | 4 | 1798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
