siRNA Details
Gene Symbol: GLTSCR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318819.1 | NP_001305748.1 | 14 | 5368 | ||
NM_015349.2 | NP_056164.1 | 12 | 5101 | |||
GenBank mRNA | AK128771.1 | - | 1 | 1230 | ||
AK294706.1 | - | 13 | 2846 | |||
AL833540.1 | - | 12 | 5101 | |||
BC047630.1 | - | 1 | 2072 | |||
CR627378.1 | - | 13 | 5066 | |||
D87077.2 | - | 15 | 5265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
