siRNA Details
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013439.2 | NP_038467.2 | 7 | 1124 | ||
NM_178272.1 | NP_840056.1 | 6 | 905 | |||
NM_178273.1 | NP_840057.1 | 5 | 871 | |||
GenBank mRNA | AF161080.1 | - | 7 | 1124 | ||
AJ400841.1 | - | 7 | 1065 | |||
AJ400842.1 | - | 6 | 822 | |||
AJ400843.1 | - | 5 | 787 | |||
BC017812.1 | - | 6 | 854 | 4690881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
