siRNA Details
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013439.2 | NP_038467.2 | 7 | 1152 | ||
NM_178272.1 | NP_840056.1 | 6 | 933 | |||
NM_178273.1 | NP_840057.1 | 5 | 899 | |||
GenBank mRNA | AJ400841.1 | - | 7 | 1093 | ||
AJ400842.1 | - | 6 | 850 | |||
AJ400843.1 | - | 5 | 815 | |||
BC017812.1 | - | 6 | 882 | 4690881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
