siRNA Details
Gene Symbol: PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178238.3 | NP_839956.1 | 2 | 562 | ||
GenBank mRNA | AF161081.1 | - | 2 | 550 | ||
AJ400845.1 | - | 2 | 570 | |||
BC050547.1 | - | 5 | 609 | 5755993 |
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013439.2 | NP_038467.2 | 2 | 474 | ||
NM_178272.1 | NP_840056.1 | 2 | 474 | |||
NM_178273.1 | NP_840057.1 | 2 | 474 | |||
GenBank mRNA | AF161080.1 | - | 2 | 474 | ||
AJ400841.1 | - | 2 | 415 | |||
AJ400842.1 | - | 2 | 391 | |||
AJ400843.1 | - | 2 | 391 | |||
AK298731.1 | - | 2 | 391 | |||
BC017812.1 | - | 2 | 461 | 4690881 |
Gene Symbol: STAG3L5P-PVRIG2P-PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036569.1 | 16 | 2807 | |||
GenBank mRNA | AJ400846.1 | - | 5 | 647 | ||
AK056412.1 | - | 16 | 2758 | |||
AK298676.1 | - | 3 | 600 | |||
AL834336.1 | - | 3 | 476 | |||
DQ851871.1 | - | 11 | 1396 | |||
DQ851877.1 | - | 9 | 1900 | |||
DQ851878.1 | - | 10 | 1489 | |||
DQ851880.1 | - | 8 | 1315 | |||
DQ851881.1 | - | 7 | 1254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
