siRNA Details
Gene Symbol: NOP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291978.1 | NP_001278907.1 | 10 | 1632 | ||
NM_001291979.1 | NP_001278908.1 | 10 | 1632 | |||
NM_003703.2 | NP_003694.1 | 10 | 1632 | |||
GenBank mRNA | AB000467.1 | - | 8 | 1028 | ||
AF040965.1 | - | 10 | 1494 | |||
AK025692.1 | - | 4 | 497 | |||
AK292939.1 | - | 10 | 1632 | |||
BC009760.2 | - | 6 | 826 | |||
BC026035.1 | - | 10 | 1459 | 4878317 | ||
BC053635.1 | - | 10 | 1511 | 5114280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
