siRNA Details
Gene Symbol: NOP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291978.1 | NP_001278907.1 | 1 | 278 | ||
NM_001291979.1 | NP_001278908.1 | 1 | 278 | |||
NM_003703.2 | NP_003694.1 | 1 | 278 | |||
GenBank mRNA | AF040965.1 | - | 1 | 140 | ||
AK292939.1 | - | 1 | 278 | |||
BC026035.1 | - | 1 | 105 | 4878317 | ||
BC053635.1 | - | 1 | 157 | 5114280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
