siRNA Details
Gene Symbol: PPP2R2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291310.1 | NP_001278239.1 | 3002 | |||
NM_018461.4 | NP_060931.2 | 1965 | ||||
NR_033191.2 | 1951 | |||||
XM_006717915.2 | XP_006717978.1 | 1381 | ||||
XM_017016406.1 | XP_016871895.1 | 2027 | ||||
XM_017016408.1 | XP_016871897.1 | 1233 | ||||
GenBank mRNA | AB040974.1 | - | 2440 | |||
AF220046.1 | - | 5 | 898 | |||
AK001483.1 | - | 2 | 601 | |||
AK293065.1 | - | 9 | 1481 | |||
BC047379.1 | - | 9 | 1965 | 5267510 | ||
BC058076.1 | - | 4 | 959 | |||
BC072402.1 | - | 6 | 1281 | 6048730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
